Pentyl-4-hydroxybenzoate 98%
Supplier: Thermo Scientific Chemicals
L05563-06
L05563-14
CAAAAL05563-06EA
70.15
CAD
CAAAAL05563-06
CAAAAL05563-14
Pentyl-4-hydroxybenzoate 98%
Pentyl 4-hydroxybenzoate
Purity: 98%
CAS No.: 6521-29-5
Molecular Formula: C12H16O3
Formula Weight: 208.26
Melting Point: 36-40°
Boiling Point: 197-200°/10mm
MDL No.: MFCD00016479
Formula:
C₁₂H₁₆O₃ Boiling Pt: 197…200 °C (10 mmHg) Melting Pt: 36…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016479 CAS Number: 6521-29-5 EINECS: 229-408-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...