2-Nitro-m-xylene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05546-22
L05546-36
CAAAAL05546-22EA
69.68
CAD
CAAAAL05546-22
CAAAAL05546-36
2-Nitro-m-xylene 99%
2-Nitro-m-xylene
Purity: 99%
CAS No.: 81-20-9
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Flash Point: 87°(188°F)
Refractive Index: 1.5220
Melting Point: 14-16°
Boiling Point: 225-227°
Beilstein Reference: 2046066
UN No.: 1665
MDL No.: MFCD00007181
Formula:
C₈H₉NO₂ Boiling Pt: 225…227 °C Melting Pt: 14…16 °C Density: 1.110 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007181 CAS Number: 81-20-9 EINECS: 201-333-6 UN: 1665 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...