4-Methyl-1-pentyne 97%

Supplier: Thermo Scientific Chemicals
Danger

L05537-03 L05537-06
CAAAAL05537-03EA 94.01 CAD
CAAAAL05537-03 CAAAAL05537-06
4-Methyl-1-pentyne 97%
4-Methyl-1-pentyne

Purity: 97% CAS No.: 7154-75-8 Molecular Formula: (CH3)2CHCH2C≡CH Formula Weight: 82.15 Flash Point: -24°(-11°F) Refractive Index: 1.3930 Boiling Point: 61-62° Beilstein Reference: 1731077 UN No.: 3295 MDL No.: MFCD00039857

Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 61…62 °C
Melting Pt: –110.3 °C
Density: 0.698
Flash Pt: –24 °C (–11 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039857
CAS Number: 7154-75-8
UN: 3295
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR