3-Butyne-2-one 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L05527-03
L05527-06
L05527-14
CAAAAL05527-03EA
104.28
CAD
CAAAAL05527-03
CAAAAL05527-06
CAAAAL05527-14
3-Butyne-2-one 98%
3-Butyne-2-one
Purity: 98%
CAS No.: 1423-60-5
Molecular Formula: C4H4O
Formula Weight: 68.08
Storage Temperature: 2°C to 8°C
Flash Point: -1°(30°F)
Refractive Index: 1.4060
Boiling Point: 84-86°
Beilstein Reference: 605353
UN No.: 1992
MDL No.: MFCD00008775
Formula:
C₄H₄O Boiling Pt: 84…86 °C Density: 0.870 Flash Pt: –1 °C (30 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008775 CAS Number: 1423-60-5 EINECS: 215-834-2 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...