Methyl palmitate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Hexadecylic acid methyl ester, Methyl hexadecanoate, Palmitic acid methyl ester, Hexadecanoic acid methyl ester, Palmitic Acid Methyl Ester

L05509-22 L05509-36
CAAAAL05509-22EA 73.79 CAD
CAAAAL05509-22 CAAAAL05509-36
Methyl palmitate 97%
Methyl palmitate

Purity: 97% CAS No.: 112-39-0 Molecular Formula: C17H34O2 Formula Weight: 270.46 Flash Point: 110°(230°F) Refractive Index: 1.4512 Melting Point: 28-34° Boiling Point: 135-137°/0.2mm Beilstein Reference: 1780973 MDL No.: MFCD00008994

Formula: C₁₇H₃₄O₂
MW: 270.46 g/mol
Boiling Pt: 417
Melting Pt: 29.9
Density: 0.852
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008994
CAS Number: 112-39-0
EINECS: 203-966-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR