1-(2-Pyridyl)piperazine 99%

Supplier: Thermo Scientific Chemicals
Warning

L05451-06 L05451-14 L05451-22
CAAAAL05451-06EA 72.36 CAD
CAAAAL05451-06 CAAAAL05451-14 CAAAAL05451-22
1-(2-Pyridyl)piperazine 99%
1-(2-Pyridyl)piperazine

Purity: 99% CAS No.: 34803-66-2 Molecular Formula: C9H13N3 Formula Weight: 163.22 Flash Point: 110°(230°F) Refractive Index: 1.5950 Boiling Point: 120-122°/2mm Beilstein Reference: 140423 UN No.: 2735 MDL No.: MFCD00006216

Formula: C₉H₁₃N₃
Boiling Pt: 120…122 °C/2 mm
Melting Pt: 85…87 °C
Density: 1.072
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006216
CAS Number: 34803-66-2
EINECS: 252-220-3
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR