1-Bromo-2-(2-methoxyethoxy)ethane tech. 90% stabilized, Technical Grade
Supplier: Thermo Scientific Chemicals
Warning
|
Stabilization:
Stabilised with sodium carbonate (Na₂CO₃)
L05406-06
L05406-14
CAAAAL05406-06EA
101.59
CAD
CAAAAL05406-06
CAAAAL05406-14
1-Bromo-2-(2-methoxyethoxy)ethane tech. 90% stabilized, Technical Grade
1-Bromo-2-(2-methoxyethoxy)ethane
Purity: 90%
CAS No.: 54149-17-6
Molecular Formula: CH3(OCH2CH2)2Br
Formula Weight: 183.05
Storage Temperature: 2°C to 8°C
Flash Point: 73°(163°F)
Refractive Index: 1.4550
Boiling Point: 156°
Beilstein Reference: 2069579
MDL No.: MFCD00000238
Formula:
BrH₂CCH₂OCH₂CH₂OCH₃ Boiling Pt: 156 °C Melting Pt: –23 °C Density: 1.347 Flash Pt: 73 °C(163 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000238 CAS Number: 54149-17-6 EINECS: 259-000-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...