Molecular sieve 3A (0.3 nm, 3 Å), beads 3-5 mm (0.12-0.20 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate

L05359-0B L05359-30
CAAAAL05359-0BEA 202.24 CAD
CAAAAL05359-0B CAAAAL05359-30
Molecular sieve 3A (0.3 nm, 3 Å), beads 3-5 mm (0.12-0.20 in)
Molecular sieve 3A (0.3 nm, 3 Å)

CAS No.: 308080-99-1 MDL No.: MFCD00131613

Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR