, beads 1-2 mm (0.04-0.08 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate

L05335-0B L05335-0I
CAAAAL05335-0BEA 188.02 CAD
CAAAAL05335-0B CAAAAL05335-0I
, beads 1-2 mm (0.04-0.08 in)
Molecular sieve 3A (0.3 nm, 3 Å)

CAS No.: 308080-99-1 MDL No.: MFCD00131613

Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR