3-Hexyne-2-one 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L05315-04
L05315-09
CAAAAL05315-04EA
114.55
CAD
CAAAAL05315-04
CAAAAL05315-09
3-Hexyne-2-one 97%
Purity: 97%
CAS No.: 1679-36-3
Molecular Formula: C6H8O
Formula Weight: 96.13
Flash Point: 39°(102°F)
Refractive Index: 1.4410
Boiling Point: 67-68°/50mm
Beilstein Reference: 1699613
UN No.: 1992
MDL No.: MFCD00041627
Formula:
C₆H₈O Boiling Pt: 67…68 °C/50 mm Density: 0.880 Flash Pt: 39 °C (102 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041627 CAS Number: 1679-36-3 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...