Methyl-3-phenylpropionate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl dihydrocinnamate, 3-Phenylpropionic acid methyl ester, Methyl 3-phenylpropionate, Hydrocinnamic acid methyl ester
L05310-18
L05310-30
CAAAAL05310-18EA
159.58
CAD
CAAAAL05310-18
CAAAAL05310-30
Methyl-3-phenylpropionate 98%
Methyl 3-phenylpropionate
Purity: 98%
CAS No.: 103-25-3
Molecular Formula: C10H12O2
Formula Weight: 164.20
Flash Point: 100°(212°F)
Refractive Index: 1.5020
Boiling Point: 236-239°
Beilstein Reference: 1909591
MDL No.: MFCD00017209
Formula:
C₁₀H₁₂O₂ MW: 164.2 g/mol Boiling Pt: 236…239 °C Density: 1,045 Flash Pt: 100 °C(212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00017209 CAS Number: 103-25-3 EINECS: 203-092-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...