α-Bromo-3,4-dichlorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L05254-06
L05254-14
CAAAAL05254-06EA
38.55
CAD
CAAAAL05254-06
CAAAAL05254-14
α-Bromo-3,4-dichlorotoluene 98%
α-Bromo-3,4-dichlorotoluene
Purity: 98%
CAS No.: 18880-04-1
Molecular Formula: C7H5BrCl2
Formula Weight: 239.93
Flash Point:
Formula:
C₇H₅BrCl₂ Boiling Pt: 116…119 °C/1 mm Density: 1.65 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041363 CAS Number: 18880-04-1 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...