4-Chloro-2,5-dimethylbenzenesulfonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Chloro-p-xylene-2-sulfonyl chloride, 4-Chloro-2,5-dimethylbenzene-1-sulfonyl chloride, 4-Chloro-2,5-dimethylbenzene-1-sulphonyl chloride, 5-Chloro-p-xylene-2-sulphonyl chloride

L05247-06 L05247-14
CAAAAL05247-06EA 87.85 CAD
CAAAAL05247-06 CAAAAL05247-14
4-Chloro-2,5-dimethylbenzenesulfonyl chloride 98%
4-Chloro-2,5-dimethylbenzenesulphonyl chloride

Purity: 98% CAS No.: 88-49-3 Molecular Formula: C8H8Cl2O2S Formula Weight: 239.12 Melting Point: 48-50° Beilstein Reference: 2838165 UN No.: 3261 MDL No.: MFCD00044017

Formula: C₈H₈Cl₂O₂S
MW: 239.12 g/mol
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00044017
CAS Number: 88-49-3
EINECS: 201-835-5
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR