2-Methylbenzylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L05243-06
L05243-14
CAAAAL05243-06EA
141.41
CAD
CAAAAL05243-06
CAAAAL05243-14
2-Methylbenzylamine 98%
2-Methylbenzylamine
Purity: 98%
CAS No.: 89-93-0
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 83°(181°F)
Refractive Index: 1.5440
Boiling Point: 198-200°
Beilstein Reference: 1099406
UN No.: 2735
MDL No.: MFCD00008112
Formula:
C₈H₁₁N Boiling Pt: 198…200 °C Melting Pt: < 0 °C Density: 0.979 Flash Pt: 83 °C (181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008112 CAS Number: 89-93-0 EINECS: 201-952-1 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...