Citraconic anhydride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Methylfuran-2,5-dione
L05238-14
L05238-22
L05238-36
CAAAAL05238-14EA
58.14
CAD
CAAAAL05238-14
CAAAAL05238-22
CAAAAL05238-36
Citraconic anhydride 98%
Citraconic anhydride
Purity: 98%
CAS No.: 616-02-4
Molecular Formula: C5H4O3
Formula Weight: 112.08
Flash Point: 101°(213°F)
Refractive Index: 1.4710
Melting Point: 7-8°
Boiling Point: 213-214°
Beilstein Reference: 1835
UN No.: 2810
MDL No.: MFCD00005522
Formula:
C₅H₄O₃ Boiling Pt: 213…214 °C Melting Pt: 7…8 °C Density: 1.243 Flash Pt: 101 °C (213 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005522 CAS Number: 616-02-4 EINECS: 210-459-0 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...