1-Iodo-3-methylbutane 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with copper

L05194-09 L05194-18
CAAAAL05194-09EA 84.21 CAD
CAAAAL05194-09 CAAAAL05194-18
1-Iodo-3-methylbutane 97% stabilized
1-Iodo-3-methylbutane

Purity: 97% CAS No.: 541-28-6 Molecular Formula: C5H11I Formula Weight: 198.05 Flash Point: 40°(104°F) Refractive Index: 1.4920 Boiling Point: 146-148° Beilstein Reference: 1730990 Merck Reference: 14,5120 UN No.: 1993 MDL No.: MFCD00039414

Formula: C₅H₁₁I
Boiling Pt: 146…148 °C
Density: 1.515
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039414
CAS Number: 541-28-6
EINECS: 208-773-8
UN: 1993
ADR: 3,III
Merck Index: 14,5120

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR