Methyl cholate 98+%
Supplier: Thermo Scientific Chemicals
L05129-06
L05129-14
CAAAAL05129-06EA
76.16
CAD
CAAAAL05129-06
CAAAAL05129-14
Methyl cholate 98+%
Methyl cholate
Purity: 98+%
CAS No.: 1448-36-8
Molecular Formula: C25H42O5
Formula Weight: 422.61
Melting Point: 155-156°
Beilstein Reference: 2226989
Merck Reference: 14,2203
MDL No.: MFCD00064934
Formula:
C₂₅H₄₂O₅ Melting Pt: 155…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064934 CAS Number: 1448-36-8 EINECS: 215-903-7 Merck Index: 14,2203 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...