2-Naphthoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: β-Naphthoic acid

L05102-06 L05102-14
CAAAAL05102-06EA 58.93 CAD
CAAAAL05102-06 CAAAAL05102-14
2-Naphthoic acid 98+%
2-Naphthoic acid

Purity: 98+% CAS No.: 93-09-4 Molecular Formula: C11H8O2 Formula Weight: 172.18 Flash Point: 205°(401°F) Melting Point: 182-186° Beilstein Reference: 972039 Merck Reference: 14,6382 MDL No.: MFCD00004101

Formula: C₁₁H₈O₂
MW: 172.18 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 182…186 °C
Density: 1.08
Flash Pt: 205 °C(401 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004101
CAS Number: 93-09-4
EINECS: 202-217-8
Merck Index: 14,6382

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR