1-Bromo-2-methylnaphthalene tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

L05093-09 L05093-18
CAAAAL05093-09EA 66.2 CAD
CAAAAL05093-09 CAAAAL05093-18
1-Bromo-2-methylnaphthalene tech. 90%, Technical Grade
1-Bromo-2-methylnaphthalene

Purity: 90% CAS No.: 2586-62-1 Molecular Formula: C11H9Br Formula Weight: 221.10 Flash Point: 110°(230°F) Refractive Index: 1.6490 Boiling Point: 295-296° Beilstein Reference: 2042531 MDL No.: MFCD00003871

Formula: C₁₁H₉Br
Boiling Pt: 295…296 °C
Density: 1.428
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003871
CAS Number: 2586-62-1
EINECS: 219-966-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR