4-Cyclohexylbenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05086-04
L05086-09
L05086-18
CAAAAL05086-04EA
76.16
CAD
CAAAAL05086-04
CAAAAL05086-09
CAAAAL05086-18
4-Cyclohexylbenzoic acid 98%
4-Cyclohexylbenzoic acid
Purity: 98%
CAS No.: 20029-52-1
Molecular Formula: C13H16O2
Formula Weight: 204.27
Melting Point: 194-198°
Beilstein Reference: 1966671
Formula:
C₁₃H₁₆O₂ Melting Pt: 194…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039459 CAS Number: 20029-52-1 EINECS: 243-472-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...