1,1-Dimethylurea 98%

Supplier: Thermo Scientific Chemicals

L05065-09 L05065-18
CAAAAL05065-09EA 92.43 CAD
CAAAAL05065-09 CAAAAL05065-18
1,1-Dimethylurea 98%
1,1-Dimethylurea

Purity: 98% CAS No.: 598-94-7 Molecular Formula: (CH3)2NCONH2 Formula Weight: 88.11 Melting Point: 178-183° Beilstein Reference: 1740666

Formula: C₃H₈N₂O
Melting Pt: 178…183 °C
Density: 1.255
Storage Temperature: Ambient
MDL Number: MFCD00007959
CAS Number: 598-94-7
EINECS: 209-957-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR