Ethyl benzoylacetate ≥90%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 3-oxo-3-phenylpropanoate

L05030-0B L05030-18 L05030-30
CAAAAL05030-0BEA 443.98 CAD
CAAAAL05030-0B CAAAAL05030-18 CAAAAL05030-30
Ethyl benzoylacetate ≥90%
Ethyl benzoylacetate

Purity: 90% CAS No.: 94-02-0 Molecular Formula: C11H12O3 Formula Weight: 192.21 Flash Point: 140°(284°F) Refractive Index: 1.5290 Boiling Point: 266-270° Beilstein Reference: 389944 Merck Reference: 14,3767 MDL No.: MFCD00009196

Formula: C₁₁H₁₂O₃
MW: 192.21 g/mol
Boiling Pt: 265
Density: 1.113
Flash Pt: 140 °C (284 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009196
CAS Number: 94-02-0
EINECS: 202-295-3
Merck Index: 14,3767

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR