2,4,6-Trimethoxybenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04997-06
L04997-14
CAAAAL04997-06EA
172.22
CAD
CAAAAL04997-06
CAAAAL04997-14
2,4,6-Trimethoxybenzoic acid 98%
2,4,6-Trimethoxybenzoic acid
Purity: 98%
CAS No.: 570-02-5
Molecular Formula: C10H12O5
Formula Weight: 212.20
Melting Point: ca 158° dec.
Beilstein Reference: 1972294
Formula:
C₁₀H₁₂O₅ Melting Pt: ca 158 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00016497 CAS Number: 570-02-5 EINECS: 209-325-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...