meso-1,2-Dibromo-1,2-diphenylethane 97%

Supplier: Thermo Scientific Chemicals
Danger

L04956-06 L04956-14
CAAAAL04956-06EA 60.83 CAD
CAAAAL04956-06 CAAAAL04956-14
meso-1,2-Dibromo-1,2-diphenylethane 97%
meso-1,2-Dibromo-1,2-diphenylethane

Purity: 97% CAS No.: 13440-24-9 Molecular Formula: C14H12Br2 Formula Weight: 340.07 Melting Point: ca 241° dec. Beilstein Reference: 2050820 UN No.: 1759 MDL No.: MFCD00000137

Formula: C₁₄H₁₂Br₂
Melting Pt: ca 241 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00000137
CAS Number: 13440-24-9
EINECS: 227-324-7
UN: 1759
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR