4-(Methylthio)aniline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Methylmercapto)aniline

L04950-06 L04950-14
CAAAAL04950-06EA 63.04 CAD
CAAAAL04950-06 CAAAAL04950-14
4-(Methylthio)aniline 98%
4-(Methylthio)aniline

Purity: 98% CAS No.: 104-96-1 Molecular Formula: C7H9NS Formula Weight: 139.23 Flash Point: 110°(230°F) Refractive Index: 1.6390 Odor: Unpleasant Boiling Point: 272-273° Beilstein Reference: 774506 UN No.: 2810 MDL No.: MFCD00007889

Formula: C₇H₉NS
MW: 139.22 g/mol
Boiling Pt: 272…273 °C
Melting Pt: 156…158 °C
Density: 1.120
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007889
CAS Number: 104-96-1
EINECS: 203-256-3
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR