(1R)-(+)-α-Pinene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(+)-α-Pinene, D(+)-α-Pinene, (1R)-(+)-2-Pinene, (+)-Pin-2(3)-ene
L04941-18
L04941-30
CAAAAL04941-18EA
71.57
CAD
CAAAAL04941-18
CAAAAL04941-30
(1R)-(+)-α-Pinene 98%
(1R)-(+)-α-Pinene
Purity: 98%
CAS No.: 7785-70-8
Molecular Formula: C10H16
Formula Weight: 136.24
Flash Point: 32°(89°F)
Refractive Index: 1.4660
Melting Point: -62°
Boiling Point: 154-156°
Beilstein Reference: 3648264
Merck Reference: 14,7445
UN No.: 2368
MDL No.: MFCD00001346
Formula:
C₁₀H₁₆ MW: 136.24 g/mol Boiling Pt: 154…156 °C Melting Pt: –62 °C Density: 0.858 Flash Pt: 32 °C (89 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001346 CAS Number: 7785-70-8 EINECS: 232-087-8 UN: 2368 ADR: 3,III Merck Index: 14,7445 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...