(±)-3-Methylcyclopentanone 99%

Supplier: Thermo Scientific Chemicals
Warning

L04940-03 L04940-06
CAAAAL04940-03EA 46.93 CAD
CAAAAL04940-03 CAAAAL04940-06
(±)-3-Methylcyclopentanone 99%
(±)-3-Methylcyclopentanone

Purity: 99% CAS No.: 1757-42-2 Molecular Formula: C6H10O Formula Weight: 98.15 Flash Point: 36°(96°F) Refractive Index: 1.4340 Melting Point: -58° Boiling Point: 144-146° Beilstein Reference: 1305074 UN No.: 1224 MDL No.: MFCD00001416

Formula: C₆H₁₀O
Boiling Pt: 144…146 °C
Melting Pt: –58 °C
Density: 0.914
Flash Pt: 36 °C (96 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001416
CAS Number: 1757-42-2
EINECS: 217-148-9
UN: 1224
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR