3-Methylbenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: m-Tolylmethanamine, 3-(Aminomethyl)toluene

L04922-03 L04922-06
CAAAAL04922-03EA 48.51 CAD
CAAAAL04922-03 CAAAAL04922-06
3-Methylbenzylamine 98%
3-Methylbenzylamine

Purity: 98% CAS No.: 100-81-2 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 80°(176°F) Refractive Index: 1.5360 Boiling Point: 203-205° Beilstein Reference: 507474 UN No.: 2735 MDL No.: MFCD00008118

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 203…205 °C
Density: 0.960
Flash Pt: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008118
CAS Number: 100-81-2
EINECS: 202-890-8
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR