1-Chloro-2-phenylethane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04891-09
L04891-22
L04891-36
CAAAAL04891-09EA
50.09
CAD
CAAAAL04891-09
CAAAAL04891-22
CAAAAL04891-36
1-Chloro-2-phenylethane 99%
1-Chloro-2-phenylethane
Purity: 99%
CAS No.: 622-24-2
Molecular Formula: C6H5(CH2)2Cl
Formula Weight: 140.61
Flash Point: 66°(150°F)
Refractive Index: 1.5300
Melting Point: -60°
Boiling Point: 82-84°/16mm
Beilstein Reference: 385969
MDL No.: MFCD00000977
Formula:
ClCH₂CH₂C₆H₅ Boiling Pt: 82…84 °C/16 mm Melting Pt: –60 °C Density: 1.070 Flash Pt: 66 °C(150 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000977 CAS Number: 622-24-2 EINECS: 210-725-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...