3-(Indol-3-yl)propionic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Indole-3-propionic acid, 3-Indolepropionic acid

L04877-06 L04877-14
CAAAAL04877-06EA 82.25 CAD
CAAAAL04877-06 CAAAAL04877-14
3-(Indol-3-yl)propionic acid 98%
3-(Indol-3-yl)propionic acid

Purity: 98% CAS No.: 830-96-6 Molecular Formula: C11H11NO2 Formula Weight: 189.21 Melting Point: 132-135° Beilstein Reference: 147733

Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00005660
CAS Number: 830-96-6
EINECS: 212-600-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR