2,3-Dibromobutyric acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L04841-06
L04841-14
CAAAAL04841-06EA
38.39
CAD
CAAAAL04841-06
CAAAAL04841-14
2,3-Dibromobutyric acid 97%
2,3-Dibromobutyric acid
Purity: 97%
CAS No.: 600-30-6
Molecular Formula: C4H6Br2O2
Formula Weight: 245.91
Melting Point: 84-88°
Boiling Point: 100-110°/2mm
UN No.: 3261
MDL No.: MFCD00014334
Formula:
H₃C(BrCH)₂COOH Boiling Pt: 100…110 °C/2 mm Melting Pt: 84…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014334 CAS Number: 600-30-6 EINECS: 209-992-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...