3-Chloro-α-methylbenzyl alcohol 97%
Supplier: Thermo Scientific Chemicals
L04783-06
L04783-14
CAAAAL04783-06EA
71.89
CAD
CAAAAL04783-06
CAAAAL04783-14
3-Chloro-α-methylbenzyl alcohol 97%
3-Chloro-α-methylbenzyl alcohol
Purity: 97%
CAS No.: 6939-95-3
Molecular Formula: C8H9ClO
Formula Weight: 156.61
Flash Point:
Formula:
C₈H₉ClO Boiling Pt: 83…85 °C/1.5 mm Density: 1,173 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00021864 CAS Number: 6939-95-3 EINECS: 230-079-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...