(Ethylthio)acetic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

L04743-06 L04743-14
CAAAAL04743-06EA 66.99 CAD
CAAAAL04743-06 CAAAAL04743-14
(Ethylthio)acetic acid 97%
(Ethylthio)acetic acid

Purity: 97% CAS No.: 627-04-3 Molecular Formula: CH3CH2SCH2CO2H Formula Weight: 120.18 Refractive Index: 1.4860 Boiling Point: 117-118°/11mm UN No.: 3265 MDL No.: MFCD00021769

Formula: C₄H₈O₂S
Boiling Pt: 117…118 °C/11 mm
Melting Pt: –8.5 °C
Density: 1.1497 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00021769
CAS Number: 627-04-3
EINECS: 210-979-8
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR