(R)-(+)-Limonene 97% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (R)-(+)-Dipentene, (R)-(+)-1-Methyl-4-(1-methylvinyl)cyclohexene, (R)-(+)-4-Isopropenyl-1-methyl-1-cyclohexene, D(+)-Limonene, (R)-(+)-p-Mentha-1,8-diene, D-Limonene

L04733-AE L04733-AP
CAAAAL04733-AEEA 47.87 CAD
CAAAAL04733-AE CAAAAL04733-AP
(R)-(+)-Limonene 97% stabilised
(R)-(+)-Limonene

Purity: 97% CAS No.: 5989-27-5 Molecular Formula: C10H16 Formula Weight: 136.23 Flash Point: 48°(118°F) Refractive Index: 1.4730 Odor: Fruit-like Melting Point: -74° Boiling Point: 177-178° Beilstein Reference: 2204754 Merck Reference: 14,5493 Optical Rotation: +115° (c=10 in ethanol) UN No.: 2052 MDL No.: MFCD00062991

Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 175.5
Melting Pt: -74
Density: 0.841
Flash Pt: 48 °C (118 °F)
Storage Temperature: Ambient
MDL Number: MFCD00062991
CAS Number: 5989-27-5
EINECS: 227-813-5
UN: 2052
ADR: 3 III
Merck Index: 14,5493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR