Bis(4-fluorophenyl) sulfone 98+%
Supplier: Thermo Scientific Chemicals
L04723-06
L04723-14
CAAAAL04723-06EA
52.77
CAD
CAAAAL04723-06
CAAAAL04723-14
Bis(4-fluorophenyl) sulfone 98+%
Bis(4-fluorophenyl) sulphone
Purity: 98+%
CAS No.: 383-29-9
Molecular Formula: C12H8F2O2S
Formula Weight: 254.25
Odor: Odorless
Melting Point: 97-101°
Beilstein Reference: 1970911
MDL No.: MFCD00000350
Formula:
C₁₂H₈F₂O₂S Melting Pt: 97…101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000350 CAS Number: 383-29-9 EINECS: 206-847-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...