Trimethylammonium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04718-18
L04718-30
CAAAAL04718-18EA
42.19
CAD
CAAAAL04718-18
CAAAAL04718-30
Trimethylammonium chloride 98%
Trimethylammonium chloride
Purity: 98%
CAS No.: 593-81-7
Molecular Formula: (CH3)3N.HCl
Formula Weight: 95.57
Melting Point: ca 280° dec.
Beilstein Reference: 3905063
Merck Reference: 14,9710
MDL No.: MFCD00012478
Formula:
C₃H₁₀ClN Melting Pt: ∼ 280 °C (dec.) Density: ~ 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00012478 CAS Number: 593-81-7 EINECS: 209-810-0 Merck Index: 14,9710 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...