4-Nitro-o-phenylenediamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Nitro-1,2-phenylenediamine, 4-Nitrobenzene-1,2-diamine

L04693-22 L04693-36
CAAAAL04693-22EA 106.02 CAD
CAAAAL04693-22 CAAAAL04693-36
4-Nitro-o-phenylenediamine 97%
4-Nitro-o-phenylenediamine

Purity: 97% CAS No.: 99-56-9 Molecular Formula: C6H7N3O2 Formula Weight: 153.14 Melting Point: 199-203° Beilstein Reference: 608106 Merck Reference: 14,6623 MDL No.: MFCD00007724

Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Melting Pt: 199…203 °C
Storage Temperature: Ambient
MDL Number: MFCD00007724
CAS Number: 99-56-9
EINECS: 202-766-3
Merck Index: 14,6623

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR