Phenyltriethoxysilane 98%

Supplier: Thermo Scientific Chemicals
Warning

L04684-0B L04684-18 L04684-30
CAAAAL04684-0BEA 224.36 CAD
CAAAAL04684-0B CAAAAL04684-18 CAAAAL04684-30
Phenyltriethoxysilane 98%
Phenyltriethoxysilane

Purity: 98% CAS No.: 780-69-8 Molecular Formula: C12H20Si Formula Weight: 240.38 Flash Point: 96°(204°F) Refractive Index: 1.4610 Boiling Point: 235° Beilstein Reference: 2940602 MDL No.: MFCD00009065

Formula: C₁₂H₂₀O₃Si
Boiling Pt: 235 °C
Melting Pt: 41 °C
Density: 0.996
Flash Pt: 96 °C (204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009065
CAS Number: 780-69-8
EINECS: 212-305-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR