4-Chlorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Chloro-4-methylbenzene
L04672-0B
L04672-22
L04672-36
CAAAAL04672-0BEA
237
CAD
CAAAAL04672-0B
CAAAAL04672-22
CAAAAL04672-36
4-Chlorotoluene 98%
4-Chlorotoluene
Purity: 98%
CAS No.: 106-43-4
Molecular Formula: C7H7Cl
Formula Weight: 126.59
Flash Point: 51°(124°F)
Refractive Index: 1.5200
Odor: Aromatic
Melting Point: 7-9°
Boiling Point: 161-163°
Beilstein Reference: 1903635
Merck Reference: 14,2171
UN No.: 2238
MDL No.: MFCD00000631
Formula:
C₇H₇Cl MW: 126.59 g/mol Boiling Pt: 161…163 °C Melting Pt: 7…9 °C Density: 1.069 Flash Pt: 51 °C (124 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000631 CAS Number: 106-43-4 EINECS: 203-397-0 UN: 2238 ADR: 3 III Merck Index: 14,2171 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...