2,3-Hexanedione 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04669-06
L04669-14
CAAAAL04669-06EA
48.51
CAD
CAAAAL04669-06
CAAAAL04669-14
2,3-Hexanedione 94%
2,3-Hexanedione
Purity: 94%
CAS No.: 3848-24-6
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 28°(82°F)
Refractive Index: 1.4120
Odor: Characteristic
Melting Point: -30°
Boiling Point: 127-128°
Beilstein Reference: 1699896
UN No.: 1224
MDL No.: MFCD00009398
Formula:
C₆H₁₀O₂ Boiling Pt: 127…128 °C Melting Pt: –30 °C Density: 0.937 Flash Pt: 28 °C (82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009398 CAS Number: 3848-24-6 EINECS: 223-350-8 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...