Dipentyl sulfide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04649-06
L04649-14
CAAAAL04649-06EA
42.82
CAD
CAAAAL04649-06
CAAAAL04649-14
Dipentyl sulfide 97%
Dipentyl sulphide
Purity: 97%
CAS No.: 872-10-6
Molecular Formula: C10H22S
Formula Weight: 174.35
Flash Point: 85°(185°F)
Refractive Index: 1.4560
Odor: Stench
Melting Point: -51°
Boiling Point: 228-230°
Beilstein Reference: 1698031
MDL No.: MFCD00027275
Formula:
C₁₀H₂₂S Boiling Pt: 228…230 °C Melting Pt: –51 °C Density: 0,840 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00027275 CAS Number: 872-10-6 EINECS: 212-820-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...