(±)-3-Methyl-3-hexanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04625-06
L04625-14
CAAAAL04625-06EA
128.14
CAD
CAAAAL04625-06
CAAAAL04625-14
(±)-3-Methyl-3-hexanol 99%
(±)-3-Methyl-3-hexanol
Purity: 99%
CAS No.: 597-96-6
Molecular Formula: C7H16O
Formula Weight: 116.20
Flash Point: 45°(113°F)
Refractive Index: 1.4235
Boiling Point: 143°
Beilstein Reference: 1719032
UN No.: 1987
MDL No.: MFCD00021839
Formula:
C₇H₁₆O Boiling Pt: 143 °C Density: 0.823 Flash Pt: 45 °C (113 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00021839 CAS Number: 597-96-6 EINECS: 209-910-4 UN: 1987 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...