1,5-Dihydroxynaphthalene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04599-0B
L04599-30
CAAAAL04599-0BEA
358.66
CAD
CAAAAL04599-0B
CAAAAL04599-30
1,5-Dihydroxynaphthalene 98%
1,5-Dihydroxynaphthalene
Purity: 98%
CAS No.: 83-56-7
Molecular Formula: C10H8O2
Formula Weight: 160.17
Flash Point: 252°(486°F)
Melting Point: ca 260° dec.
Beilstein Reference: 2044951
MDL No.: MFCD00003980
Formula:
C₁₀H₈O₂ Boiling Pt: 375 °C (1013 hPa) Melting Pt: ∼ 260 °C (dec.) Density: 1.41 g/cm³ (20 °C) Flash Pt: 252 °C (486 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003980 CAS Number: 83-56-7 EINECS: 201-487-4 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...