(±)-1-Phenoxy-2-propanol tech. 85%, Technical Grade
Supplier: Thermo Scientific Chemicals
Synonyms:
1-Phenoxy-2-propanol, Propylene Phenoxytol, (±)-Monopropylene glycol phenyl ether
L04596-18
L04596-30
CAAAAL04596-18EA
62.41
CAD
CAAAAL04596-18
CAAAAL04596-30
(±)-1-Phenoxy-2-propanol tech. 85%, Technical Grade
(±)-1-Phenoxy-2-propanol
Purity: 85%
CAS No.: 770-35-4
Molecular Formula: C9H12O2
Formula Weight: 152.19
Flash Point: 120°(248°F)
Refractive Index: 1.5230
Boiling Point: 242-244°
Beilstein Reference: 1941356
MDL No.: MFCD00016861
Formula:
C₆H₅OCH₂CH(OH)CH₃ MW: 152.19 g/mol Boiling Pt: 242…244 °C Melting Pt: 11,4 °C Density: 1.060 Flash Pt: 120 °C(248 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00016861 CAS Number: 770-35-4 EINECS: 212-222-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...