2-Chloro-1,3-dinitrobenzene 98+%

Supplier: Thermo Scientific Chemicals
Danger

L04593-03 L04593-06
CAAAAL04593-03EA 98.28 CAD
CAAAAL04593-03 CAAAAL04593-06
2-Chloro-1,3-dinitrobenzene 98+%
2-Chloro-1,3-dinitrobenzene

Purity: 98+% CAS No.: 606-21-3 Molecular Formula: C6H3ClN2O4 Formula Weight: 202.55 Melting Point: 83-88° Boiling Point: 315° Merck Reference: 14,2137 UN No.: 3441 MDL No.: MFCD00014689

Formula: C₆H₃ClN₂O₄
Boiling Pt: 315 °C
Melting Pt: 83…88 °C
Density: 1.687
Storage Temperature: Ambient
MDL Number: MFCD00014689
CAS Number: 606-21-3
EINECS: 210-107-6
UN: 3441
ADR: 6.1,II
Merck Index: 14,2137

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR