N-(3-Hydroxypropyl)phthalimide 98%

Supplier: Thermo Scientific Chemicals

L04507-09 L04507-18
CAAAAL04507-09EA 44.56 CAD
CAAAAL04507-09 CAAAAL04507-18
N-(3-Hydroxypropyl)phthalimide 98%
N-(3-Hydroxypropyl)phthalimide

Purity: 98% CAS No.: 883-44-3 Molecular Formula: C11H11NO3 Formula Weight: 205.22 Melting Point: 75-80° Beilstein Reference: 158489

Formula: C₁₁H₁₁NO₃
Melting Pt: 75…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00023097
CAS Number: 883-44-3
EINECS: 212-931-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR