N,N-Diethylformamide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04503-22
L04503-36
CAAAAL04503-22EA
139.83
CAD
CAAAAL04503-22
CAAAAL04503-36
N,N-Diethylformamide 99%
N,N-Diethylformamide
Purity: 99%
CAS No.: 617-84-5
Molecular Formula: HCON(CH2CH3)2
Formula Weight: 101.15
Flash Point: 65°(149°F)
Refractive Index: 1.4340
Odor: Amine-like
Boiling Point: 178°
Beilstein Reference: 1209392
MDL No.: MFCD00003287
Formula:
C₅H₁₁NO Boiling Pt: 178 °C Density: 0.910 Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003287 CAS Number: 617-84-5 EINECS: 210-533-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...