1,2,4,5-Tetramethylbenzene 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L04493-22
L04493-36
CAAAAL04493-22EA
53.72
CAD
CAAAAL04493-22
CAAAAL04493-36
1,2,4,5-Tetramethylbenzene 97+%
1,2,4,5-Tetramethylbenzene
Purity: 97+%
CAS No.: 95-93-2
Molecular Formula: C10H14
Formula Weight: 134.22
Flash Point: 73°(163°F)
Melting Point: 77-80°
Boiling Point: 191-193°
Beilstein Reference: 1903393
Merck Reference: 14,3468
UN No.: 1325
MDL No.: MFCD00008528
Formula:
C₁₀H₁₄ MW: 134.22 g/mol Boiling Pt: 191…193 °C Melting Pt: 77…80 °C Density: 1.036 g/cm³ (20 °C) Flash Pt: 73 °C (163 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008528 CAS Number: 95-93-2 EINECS: 202-465-7 UN: 1325 ADR: 4.1,III Merck Index: 14,3468 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...