1,1,2,2-Tetrachloroethane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L04489-14
L04489-22
L04489-36
CAAAAL04489-14EA
55.62
CAD
CAAAAL04489-14
CAAAAL04489-22
CAAAAL04489-36
1,1,2,2-Tetrachloroethane 98+%
1,1,2,2-Tetrachloroethane
Purity: 98+%
CAS No.: 79-34-5
Molecular Formula: C2H2Cl4
Formula Weight: 167.85
Refractive Index: 1.4935
Melting Point: -43°
Boiling Point: 142-146°
Beilstein Reference: 969206
Merck Reference: 14,9189
UN No.: 1702
MDL No.: MFCD00000848
Formula:
Cl₂HCCHCl₂ Boiling Pt: 142…146 °C Melting Pt: –43 °C Density: 1.596 Storage Temperature: Ambient |
MDL Number:
MFCD00000848 CAS Number: 79-34-5 EINECS: 201-197-8 UN: 1702 ADR: 6.1,II Merck Index: 14,9189 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...