2-Picoline-N-oxide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L04488-14
L04488-22
L04488-36
CAAAAL04488-14EA
49.14
CAD
CAAAAL04488-14
CAAAAL04488-22
CAAAAL04488-36
2-Picoline-N-oxide 98%
2-Picoline N-oxide
Purity: 98%
CAS No.: 931-19-1
Molecular Formula: C6H7NO
Formula Weight: 109.13
Flash Point: 143°(289°F)
Melting Point: 46-50°
Boiling Point: 122°/8mm
Beilstein Reference: 106916
MDL No.: MFCD00006199
Formula:
C₆H₇NO Boiling Pt: 122 °C/8 mm Melting Pt: 46…50 °C Flash Pt: 143 °C(289 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006199 CAS Number: 931-19-1 EINECS: 213-230-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...